Tisk…

2-(Allyloxy)-4-methoxybenzyl chloride

Dodavatel: Apollo Scientific

APOSOR470703-1GEA 26090 CZK
APOSOR470703-1G
2-(Allyloxy)-4-methoxybenzyl chloride
2-(Allyloxy)-4-methoxybenzyl chloride
Vzorec: C₁₁H₁₃ClO₂
MW: 212,673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26937559
CAS číslo: 1823868-00-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR