2'-(Allyloxy)acetophenone
Dodavatel: Apollo Scientific
APOSOR470717-5GEA
17130
CZK
APOSOR470717-5G
2'-(Allyloxy)acetophenone
2'-(Allyloxy)acetophenone
Vzorec:
C₁₁H₁₂O₂ MW: 176,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00026218 CAS číslo: 53327-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...