2-(2-Methyl-1-piperazinyl)acetic acid
Dodavatel: Apollo Scientific
APOSOR470718-1GEA
26090
CZK
APOSOR470718-1G
2-(2-Methyl-1-piperazinyl)acetic acid
2-(2-Methyl-1-piperazinyl)acetic acid
Vzorec:
C₇H₁₄N₂O₂ MW: 158,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10698344 CAS číslo: 1060813-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...