(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid
Dodavatel: Apollo Scientific
APOSOR470728-1GEA
20300
CZK
APOSOR470728-1G
(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid
(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid
Vzorec:
C₈H₈O₃ MW: 152,147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08310685 CAS číslo: 71155-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...