2-(Allyloxy)-4-methoxybenzyl Bromide
Dodavatel: Apollo Scientific
APOSOR470730-1GEA
26090
CZK
APOSOR470730-1G
2-(Allyloxy)-4-methoxybenzyl Bromide
2-(Allyloxy)-4-methoxybenzyl Bromide
Vzorec:
C₁₁H₁₃BrO₂ MW: 257,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26937561 CAS číslo: 1823942-82-0 |