Tisk…

Benzofuran-7-carbonitrile

Dodavatel: Apollo Scientific

APOSOR470745-1GEA 20300 CZK
APOSOR470745-1G
Benzofuran-7-carbonitrile
Benzofuran-7-carbonitrile
Vzorec: C₉H₅NO
MW: 143,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143950
CAS číslo: 52951-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR