Tisk…

4-Chloro-N,N-diethyl-1-butanamine hydrochloride

Dodavatel: Apollo Scientific

APOSOR470749-1GEA 17130 CZK
APOSOR470749-1G
4-Chloro-N,N-diethyl-1-butanamine hydrochloride
4-Chloro-N,N-diethyl-1-butanamine hydrochloride
Vzorec: C₈H₁₈ClN·ClH
MW: 200,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27500641
CAS číslo: 108130-45-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR