Tisk…

Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate

Dodavatel: Apollo Scientific

APOSOR470771-5GEA 17130 CZK
APOSOR470771-5G
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate
Vzorec: C₁₂H₁₀BrNO₂
MW: 280,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00176211
CAS číslo: 18861-58-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR