Tisk…

2-Chloro-3-phenylpyrazine

Dodavatel: Apollo Scientific

APOSOR470777-1GEA 31910 CZK
APOSOR470777-1G
2-Chloro-3-phenylpyrazine
2-Chloro-3-phenylpyrazine
Vzorec: C₁₀H₇ClN₂
MW: 190,629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01411765
CAS číslo: 41270-65-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR