Tisk…

2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid

Dodavatel: Apollo Scientific

APOSOR470792-5GEA 20300 CZK
APOSOR470792-5G
2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid
2-[3-(Benzyloxy)-4-methoxyphenyl]acetic acid
Vzorec: C₁₆H₁₆O₄
MW: 272,296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00183334
CAS číslo: 5487-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR