Tisk…

tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate

Dodavatel: Apollo Scientific

APOSOR470802-1GEA 22920 CZK
APOSOR470802-1G
tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate
tert-Butyl (S)-3-(Boc-amino)-4-bromobutanoate
Vzorec: C₁₃H₂₄BrNO₄
MW: 338,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27500656
CAS číslo: 2006286-94-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR