Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate
Dodavatel: Apollo Scientific
APOSOR470822-1GEA
11310
CZK
APOSOR470822-1G
Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate
Methyl 5-[4-(ethoxycarbonyl)phenyl]isoxazole-3-carboxylate
Vzorec:
C₁₄H₁₃NO₅ MW: 275,257 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27935378 CAS číslo: 2006277-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...