Tisk…

2-Benzyl-2,6-diazaspiro[4.5]decane

Dodavatel: Apollo Scientific

APOSOR470849-1GEA 40340 CZK
APOSOR470849-1G
2-Benzyl-2,6-diazaspiro[4.5]decane
2-Benzyl-2,6-diazaspiro[4.5]decane
Vzorec: C₁₅H₂₂N₂
MW: 230,349 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223525
CAS číslo: 1086395-73-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR