Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate
Dodavatel: Apollo Scientific
APOSOR470860-5GEA
20300
CZK
APOSOR470860-5G
Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate
Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate
Vzorec:
C₈H₁₁NO₆S₃ MW: 313,371 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26381896 CAS číslo: 214532-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...