Tisk…

2-(Boc-amino)-1-iodopropane

Dodavatel: Apollo Scientific

APOSOR470869-5GEA 28700 CZK
APOSOR470869-5G
2-(Boc-amino)-1-iodopropane
2-(Boc-amino)-1-iodopropane
Vzorec: C₈H₁₆INO₂
MW: 285,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17214622
CAS číslo: 363599-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR