6-Chloro-4-hydroxy-2-(methylthio)pyrimidine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSOR470872-5GEA
20300
CZK
APOSOR470872-5G
6-Chloro-4-hydroxy-2-(methylthio)pyrimidine
6-Chloro-4-hydroxy-2-(methylthio)pyrimidine
Vzorec:
C₅H₅ClN₂OS MW: 176,624 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16875711 CAS číslo: 6632-63-9 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...