Tisk…

5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester

Dodavatel: Apollo Scientific

APOSOR470887-1GEA 43640 CZK
APOSOR470887-1G
5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester
5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester
Vzorec: C₁₄H₂₂BNO₂
MW: 247,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18730174
CAS číslo: 1011460-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR