2-[2-(Methylsulphonyl)phenyl]acetic acid
Dodavatel: Apollo Scientific
APOSOR470917-5GEA
37690
CZK
APOSOR470917-5G
2-[2-(Methylsulphonyl)phenyl]acetic acid
2-[2-(Methylsulphonyl)phenyl]acetic acid
Vzorec:
C₉H₁₀O₄S MW: 214,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11898924 CAS číslo: 142336-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...