4-Ethylpyridine-3-boronic acid pinacol ester
Dodavatel: Apollo Scientific
APOSOR470927-1GEA
36040
CZK
APOSOR470927-1G
4-Ethylpyridine-3-boronic acid pinacol ester
4-Ethylpyridine-3-boronic acid pinacol ester
Vzorec:
C₁₃H₂₀BNO₂ MW: 233,114 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13182290 CAS číslo: 2096330-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...