Ethyl 2-sulphamoylacetate
Dodavatel: Apollo Scientific
APOSOR470950-1GEA
22920
CZK
APOSOR470950-1G
Ethyl 2-sulphamoylacetate
Ethyl 2-sulphamoylacetate
Vzorec:
C₄H₉NO₄S MW: 167,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12783410 CAS číslo: 55897-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...