2-[(1,2,4-Triazol-1-yl)methyl]aniline
Dodavatel: Apollo Scientific
APOSOR470961-5GEA
17130
CZK
APOSOR470961-5G
2-[(1,2,4-Triazol-1-yl)methyl]aniline
2-[(1,2,4-Triazol-1-yl)methyl]aniline
Vzorec:
C₉H₁₀N₄ MW: 174,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025845 CAS číslo: 127988-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...