Tisk…

Methyl 6-(Isobutylamino)pyridazine-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR470967-1GEA 14220 CZK
APOSOR470967-1G
Methyl 6-(Isobutylamino)pyridazine-3-carboxylate
Methyl 6-(Isobutylamino)pyridazine-3-carboxylate
Vzorec: C₁₀H₁₅N₃O₂
MW: 209,245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12152728
CAS číslo: 1184230-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR