Tisk…

4-[(Cyclopropylamino)sulphonyl]benzoic acid

Dodavatel: Apollo Scientific

APOSOR470987-5GEA 20300 CZK
APOSOR470987-5G
4-[(Cyclopropylamino)sulphonyl]benzoic acid
4-[(Cyclopropylamino)sulphonyl]benzoic acid
Vzorec: C₁₀H₁₁NO₄S
MW: 241,264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035198
CAS číslo: 436092-71-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR