3-Amino-3-methyl-1-butanol hydrochloride
Dodavatel: Apollo Scientific
APOSOR471003-5GEA
22920
CZK
APOSOR471003-5G
3-Amino-3-methyl-1-butanol hydrochloride
3-Amino-3-methyl-1-butanol hydrochloride
Vzorec:
C₅H₁₃NO·ClH MW: 139,624 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26407724 CAS číslo: 357185-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...