Methyl 3-Amino-2-hydroxypropanoate hydrochloride
Dodavatel: Apollo Scientific
APOSOR471011-1GEA
2280
CZK
APOSOR471011-1G
Methyl 3-Amino-2-hydroxypropanoate hydrochloride
Methyl 3-Amino-2-hydroxypropanoate hydrochloride
Vzorec:
C₄H₉NO₃·ClH MW: 155,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22741315 CAS číslo: 186393-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...