Tisk…

Methyl N-Boc-3-amino-2-hydroxypropanoate

Dodavatel: Apollo Scientific

APOSOR471013-1GEA 28700 CZK
APOSOR471013-1G
Methyl N-Boc-3-amino-2-hydroxypropanoate
Methyl N-Boc-3-amino-2-hydroxypropanoate
Vzorec: C₉H₁₇NO₅
MW: 219,235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447448
CAS číslo: 113525-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR