Tisk…

4-Amino-1-methylpyrazole-3-carboxylic acid

Dodavatel: Apollo Scientific

APOSOR471022-1GEA 11080 CZK
APOSOR471022-1G
4-Amino-1-methylpyrazole-3-carboxylic acid
4-Amino-1-methylpyrazole-3-carboxylic acid
Vzorec: C₅H₇N₃O₂
MW: 141,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08558492
CAS číslo: 1006334-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR