Tisk…

Methyl (R)-2-(3-Piperidyl)acetate hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

APOSOR471028-1GEA 8800 CZK
APOSOR471028-1G
Methyl (R)-2-(3-Piperidyl)acetate hydrochloride 95%
Methyl (R)-2-(3-Piperidyl)acetate hydrochloride
Vzorec: C₈H₁₅NO₂·ClH
MW: 193,671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094723
CAS číslo: 865157-03-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR