Tisk…

1-Cyclopentylimidazole

Dodavatel: Apollo Scientific

APOSOR471036-5GEA 37690 CZK
APOSOR471036-5G
1-Cyclopentylimidazole
1-Cyclopentylimidazole
Vzorec: C₈H₁₂N₂
MW: 136,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18889431
CAS číslo: 71614-58-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR