Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate
Dodavatel: Apollo Scientific
APOSOR471077-1GEA
12770
CZK
APOSOR471077-1G
Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate
Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate
Vzorec:
C₁₅H₁₉NO₄ MW: 277,316 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23701515 CAS číslo: 141870-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...