Tisk…

1-Nitro-2-propoxybenzene

Dodavatel: Apollo Scientific

APOSOR471084-5GEA 20300 CZK
APOSOR471084-5G
1-Nitro-2-propoxybenzene
1-Nitro-2-propoxybenzene
Vzorec: C₉H₁₁NO₃
MW: 181,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122812
CAS číslo: 3079-53-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR