2-(Cbz-amino)-2-(2-pyridyl)acetic acid
Dodavatel: Apollo Scientific
APOSOR471114-5GEA
37690
CZK
APOSOR471114-5G
2-(Cbz-amino)-2-(2-pyridyl)acetic acid
2-(Cbz-amino)-2-(2-pyridyl)acetic acid
Vzorec:
C₁₅H₁₄N₂O₄ MW: 286,283 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28144003 CAS číslo: 1822467-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...