Tisk…

3-(3-Hydroxyphenyl)butanoic acid

Dodavatel: Apollo Scientific

APOSOR471133-1GEA 16240 CZK
APOSOR471133-1G
3-(3-Hydroxyphenyl)butanoic acid
3-(3-Hydroxyphenyl)butanoic acid
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20643753
CAS číslo: 51038-69-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR