Tisk…

N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide

Dodavatel: Apollo Scientific

APOSOR471134-5GEA 17130 CZK
APOSOR471134-5G
N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide
N-[4-(3-Nitrophenyl)-2-thiazolyl]acetamide
Vzorec: C₁₁H₉N₃O₃S
MW: 263,272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01023820
CAS číslo: 53173-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR