Tisk…

Methyl 2-(3-Oxocyclohexyl)acetate

Dodavatel: Apollo Scientific

Varování

APOSOR471138-1GEA 15080 CZK
APOSOR471138-1G
Methyl 2-(3-Oxocyclohexyl)acetate
Methyl 2-(3-Oxocyclohexyl)acetate
Vzorec: C₉H₁₄O₃
MW: 170,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00154577
CAS číslo: 2808-12-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR