Tisk…

2-Methyl-3-(2-nitrophenyl)propanoic acid

Dodavatel: Apollo Scientific

APOSOR471140-1GEA 12200 CZK
APOSOR471140-1G
2-Methyl-3-(2-nitrophenyl)propanoic acid
2-Methyl-3-(2-nitrophenyl)propanoic acid
Vzorec: C₁₀H₁₁NO₄
MW: 209,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12794488
CAS číslo: 60031-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR