Tisk…

tert-Butyl 2-(2-Pyrrolyl)acetate

Dodavatel: Apollo Scientific

APOSOR471142-1GEA 11310 CZK
APOSOR471142-1G
tert-Butyl 2-(2-Pyrrolyl)acetate
tert-Butyl 2-(2-Pyrrolyl)acetate
Vzorec: C₁₀H₁₅NO₂
MW: 181,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20662108
CAS číslo: 157071-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR