Methyl 2-(4-Vinylphenyl)acetate
Dodavatel: Apollo Scientific
APOSOR471170-1GEA
17130
CZK
APOSOR471170-1G
Methyl 2-(4-Vinylphenyl)acetate
Methyl 2-(4-Vinylphenyl)acetate
Vzorec:
C₁₁H₁₂O₂ MW: 176,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18431776 CAS číslo: 62667-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...