Tisk…

4-Benzoyl-5-methyldihydrofuran-2(3H)-one

Dodavatel: Apollo Scientific

APOSOR471174-1GEA 12770 CZK
APOSOR471174-1G
4-Benzoyl-5-methyldihydrofuran-2(3H)-one
4-Benzoyl-5-methyldihydrofuran-2(3H)-one
Vzorec: C₁₂H₁₂O₃
MW: 204,222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348658
CAS číslo: 101563-78-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR