Tisk…

Ethyl 4-(Boc-aminomethyl)benzoate

Dodavatel: Apollo Scientific

APOSOR471182-5GEA 17130 CZK
APOSOR471182-5G
Ethyl 4-(Boc-aminomethyl)benzoate
Ethyl 4-(Boc-aminomethyl)benzoate
Vzorec: C₁₅H₂₁NO₄
MW: 279,332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10609786
CAS číslo: 157311-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR