1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide
Dodavatel: Apollo Scientific
APOSOR471187-1GEA
11310
CZK
APOSOR471187-1G
1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide
1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide
Vzorec:
C₈H₁₇BrN₂·2BrH MW: 382,962 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03452742 CAS číslo: 5845-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...