2,6-Dichloroquinazolin-4(3H)-one
Dodavatel: Apollo Scientific
APOSOR471282-5GEA
41330
CZK
APOSOR471282-5G
2,6-Dichloroquinazolin-4(3H)-one
2,6-Dichloroquinazolin-4(3H)-one
Vzorec:
C₈H₄Cl₂N₂O MW: 215,036 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520689 CAS číslo: 20197-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...