Tisk…

2-(Hydroxymethyl)-5-iodobenzimidazole

Dodavatel: Apollo Scientific

APOSOR471360-1GEA 28700 CZK
APOSOR471360-1G
2-(Hydroxymethyl)-5-iodobenzimidazole
2-(Hydroxymethyl)-5-iodobenzimidazole
Vzorec: C₈H₇IN₂O
MW: 274,058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00457490
CAS číslo: 1935632-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR