4,7-Dichloro-3-nitroquinoline
Dodavatel: Apollo Scientific
APOSOR471385-1GEA
51910
CZK
APOSOR471385-1G
4,7-Dichloro-3-nitroquinoline
4,7-Dichloro-3-nitroquinoline
Vzorec:
C₉H₄Cl₂N₂O₂ MW: 243,046 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17170308 CAS číslo: 22931-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...