4-Chloro-5-nitroquinazoline
Dodavatel: Apollo Scientific
APOSOR471386-1GEA
37690
CZK
APOSOR471386-1G
4-Chloro-5-nitroquinazoline
4-Chloro-5-nitroquinazoline
Vzorec:
C₈H₄ClN₃O₂ MW: 209,589 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18448299 CAS číslo: 75224-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...