Tisk…

3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid

Dodavatel: Apollo Scientific

APOSOR471404-1GEA 34720 CZK
APOSOR471404-1G
3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Vzorec: C₁₆H₁₆BrNO₅
MW: 382,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977386
CAS číslo: 2006277-37-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR