Tisk…

Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate

Dodavatel: Apollo Scientific

APOSOR471410-1GEA 34720 CZK
APOSOR471410-1G
Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Vzorec: C₁₃H₁₃NO₄
MW: 247,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977380
CAS číslo: 2006277-85-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR