Tisk…

(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid

Dodavatel: Apollo Scientific

APOSOR471421-1GEA 22920 CZK
APOSOR471421-1G
(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Vzorec: C₁₀H₁₉NO₂
MW: 185,263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20656288
CAS číslo: 753435-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR