4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Dodavatel: Apollo Scientific
APOSOR471428-1GEA
22920
CZK
APOSOR471428-1G
4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Vzorec:
C₁₀H₁₉NO₂ MW: 185,263 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09035339 CAS číslo: 937669-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...