Tisk…

4-Guanidino-2-methoxybenzoic acid hydrochloride

Dodavatel: Apollo Scientific

APOSOR471480-1GEA 46290 CZK
APOSOR471480-1G
4-Guanidino-2-methoxybenzoic acid hydrochloride
4-Guanidino-2-methoxybenzoic acid hydrochloride
Vzorec: C₉H₁₁N₃O₃·ClH
MW: 245,663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29035074
CAS číslo: 1028486-08-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR